Learn More
Bioworld Lens culinaris (Lentil) Lectin (LCA/LCH) 5mg
Supplier: Bioworld 217610352
Technical Specifications:Ligand Source:Lens culinarisMolecular Weight:50 kDaCarbohydrate Specificity:αMan (High Mannose), Fucα6GlcNAcInhibiting/Eluting Sugar:a-Methylglucoside +a-MethylmannosideCarbohydrate for Elution:0.2Ma-Methylglucoside +a-MethylmannosideBlood Group Specificity:N/AConjugate: Texas RedForm:LiquidIsoelectric (pI):7.6-8.4Storage: -20°CApplication note: Calcium and Manganese are required for binding.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.