Learn More
Electron Microscopy Sciences Lembert Bone Rongeur, Slightly Curved
Supplier: Electron Microscopy Sciences 77982C
Bone Rongeur. The jaw width is 2.5 mm and the length is 6 1/2". Made from stainless steel with a polished finish. Slightly curved.