Learn More
Electron Microscopy Sciences Leit-C-Plast 15 GR
Supplier: Electron Microscopy Sciences 12667
A special adhesive with very high electrical conductivity and permanent plasticity for the preparation of big specimens in SEM work.