Learn More
Electron Microscopy Sciences Applied Physics CeB6 Cathode for Leica Lithography MVM
Supplier: Electron Microscopy Sciences 80929CM
Lanthanum hexaboride (LaB6) and cerium hexaboride (CeB6) cathodes are ideal for many small spot size applications such as SEM, TEM, surface analysis and metrology, and for high current applications such as microwave tubes, lithography, electron-beam welders, X-ray sources and free electron lasers. Greater brightness from low work function yielding higher currents at lower temps than tungsten. Cambridge MVM base.
Type: CEBIX (CeB6)
Angle: 90°
Tips: 20 mm
Type: non-shunted
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.