Learn More
Coast™ LED145 LED Spring-Loaded Pro Pocket Pliers
With 15 tools, the LED145 Multi-Tool will make you prepared to tackle just about any challenge that arises.
Supplier: Coast™ C5799B
Includes: Sheath
Description
With 15 tools, the LED145 Multi-Tool will make you prepared to tackle just about any challenge that arises. Perfect to carry on your belt with a sheath, in your bag or your toolbox.
Tools Included:
Spring-loaded pliers with wire cutter, 1.75 in Blade, built-in LED, scissors, saw blade, Phillips (1) and flathead (3) screwdriver tips, bottle opener, awl, file blade with wire stripper, can opener. 2 x CR1220 batteries included.
With Coast’s commitment to quality, the LED145 Multi-Tool is backed by Coast’s lifetime warranty against defects in materials and workmanship.
Certifications
ANSI/FL1 Standards
Compliance
ANSI/FL1 Standards
Warranty and Services
Lifetime
Specifications
Multi-Tool | |
Stainless Steel | |
Multi-tool | |
0.48 kg |
Black | |
8.7 cm | |
8.7 cm | |
14 cm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.