Learn More
Caron Products And Services LED Plant Growth, 1-Door, 22 cu. ft., 1500 mole, 1 Tier
Supplier: Caron Products And Services 7317221
LED Plant Growth Chamber optimized for medicinal plants. 22 ft3 with 1500 mole/m2/s light, 1 tier, temperature control, touchscreen. 10C to 45C, 115V, 60 Hz.