Learn More
Research Products International Corp LED Digital See-Saw Rocker
Supplier: Research Products International Corp 400704
LED Digital See-Saw Rocker provides perfect motion for cell culture, sample washing, molecular hybridization, immunoprocipitation, Western blotting, and electrophoresis gel dyeing. Rocker features uniform rocking motion (7°) at speed of 10-80rpm. Suitable for mixing liquids in a variety of vessels including micro-tubes, Petri dishes, cell culture flasks, Erlenmeyer flasks, tubes, microplates and more. Unit features speed and timer LED for independent display of time and speed; continuous or timed operation and straps to secure items. Brushless motor offers maximum longevity, stable operation and low vibration.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.