Learn More
Starplex™ Scientific Leakbuster™ Containers
For collection and transportation of specimens
Supplier: Starplex™ Scientific WB9021OTS
Description
- Stable, stackable straight-sided containers
- With O-Ring cap to grip top and sides of container
- With QuickTurn™ cap for widemouth containers w/double-thread design
- Available in sterile and non-sterile formats
Specifications
Sterile | |
Orange | |
400/Cs. |
90 mL | |
Wide-mouth |