Learn More
Andwin Scientific LEAD DONUT C SHAPE 500-2000ML
Supplier: Andwin Scientific EF8880C
The lead ring is vinyl-coated and in C-Shape. It can work with flasks with volume capacity of 500 to 2000 mL. This product allows flasks to be immersed in water baths, lessening risks of breakage and spills.