Learn More
Cole-Parmer Transfer Pipette, 7.5 mL, Large Bulb, Graduated to 3 mL; 500/Box
Supplier: Cole-Parmer 0622601
Argos Technologies Transfer Pipette, 7.5 mL, Large Bulb, Graduated to 3 mL; 500/Box, No more breakage or cross-contaminationTranslucent shatterproof LDPE is nontoxic and biologically inertNonbinding for cells and proteins to prevent sample lossHeat sealable and liquid nitrogen compatibleLarge bulb pipette has a length of 155 mm and 0.5 mL graduation incrimentsDesigned with a 3.2 µL bulb draw and 20±1 drops per mL
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.