Learn More
LC Multiresidue Pesticide Kit, Restek
![SDP](/content/dam/fishersci/glyphs/sdp.png)
For detecting and quantifying pesticides of global food safety concern in a range of fruits, vegetables, and other commodities by LC-MS/MS
Supplier: Restek Corporation 31971
Description
Kit contains 204 compounds of interest, covering many LC-determined pesticides listed by government agencies
- Individual ampuls also sold separately
- Formulated and grouped for max long-term stability
- Quantitatively tested to confirm composition (support docs provided)
- Optimizes multiresidue pesticide method offered free of charge
- Downloadable XLS file includes conditions and transition tables
Notes:
- When combining a large number of compounds with different chemical functionalities, mix stability can be an issue
- When formulating standards, 204 compounds were extensively studied and grouped into as few mixes as possible while ensuring max long-term stability and reliability
- For quantitative analysis, it is recommended to analyze each mix separately to ensure accurate results for every compound
Specifications
LC-MS/MS | |
Certified reference material | |
Ambient |
Safety and Handling
ShelfLife : 6 months min on ship date
Recommended Storage : Store at -20°C or colder