Learn More
LABSYSTEMS DIAGNOSTICS OY LC-MSMS kit for CAH diagnosis
Supplier: LABSYSTEMS DIAGNOSTICS OY 7150022
This NeoMass CAH kit is intended for the determination of Androstenedione (ANDRO), Cortisol (CORT), 11-Deoxycortisol (11-DOC), 21-Deoxycortisol (21-DOC), 17-Hydroxyprogesterone (17-OHP) & 21-Hydroxyprogesterone (21-OHP) from whole blood dried on ¬filter paper as a second tier test for detection of Congenital Adrenal Hyperplasia (CAH) in newborns. The measurement of specific steroids with NeoMass CAH kit requires the extraction of dried blood spot samples from newborns with a solution containing stable isotope labeled internal standards (ISTDs) followed by concentration of the extracted solution using sample concentrator, then reconstitute with suitable solvent. The extracted samples are analyzed in LC-MSMS.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.