Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents™ LB Broth, Miller (Pre-Buffered Capsules)
Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.
Description
- Directions for Media Preparation: Add 1 capsule per 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve capsule. Before use, sterilize by autoclaving for 15 minutes.
- Product is in capsule form
- Each capsule contains 1g of broth
- Capsules are prebuffered and set to pH 7.2 at 20°C using a 10mM solution of tris/tris HCl
- pH will vary with temperature
Specifications
Pre-Buffered Capsules | |
Microbiology Media | |
For maintenance and propagation of Escherichia coli in molecular biology procedures | |
500 x 1 g |
LB Broth | |
Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L, TRIS/TRIS HCl 1.5g/L | |
Poly Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction