Learn More
United States Biological Corporation LB Broth Lennox, Powder, 2.5 kg
Supplier: United States Biological Corporation L15052.5KG
LB Lennox, a nutritionally rich medium, was developed by Lennox for the growth and maintenance of pure cultures of recombinant strains of E. coli. These strains are generally derived from E. coli K12 which are deficient in B vitamin production. LB Broth media formulations have been industry standards for the cultivation of Escherichia coli since the 1950’s. These media have been widely used in molecular microbiology applications for the preparation of plasmid DNA and recombinant proteins.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.