Learn More
Hach Company Lauryl Tryptose (with MUG) Broth Tubes, concentrated, pk/15
Supplier: Hach Company 2182115
Lauryl Tryptose Broth with MUG provides a selective medium which is used for enumeration of E. coli and other coliform bacteria in water using the Most Probable Number (MPN) method.