Learn More
United Scientific™ Lattice Lab Replacement Parts
Lattice Lab Replacement Parts allow all sets to be customized or expanded.
$4.55 - $11.25
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S43734
|
United Scientific™
LLSRC2 |
Rod Connector |
Each for $11.25
|
|
|||||
S43735
|
United Scientific™
LLSCLMP |
Hook Connector |
Each for $5.40
|
|
|||||
S43736
|
United Scientific™
LLSFPS |
Foot Plate |
Each for $4.55
|
|
|||||
S43737
|
United Scientific™
LLSR02 |
Aluminum Rod, 2 in. |
Each for $2.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S43738
|
United Scientific™
LLSR24 |
Aluminum Rod, 24 in. |
Each for $7.50
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S43739
|
United Scientific™
LLSR36 |
Aluminum Rod, 36 in. |
Each for $11.25
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S43740
|
United Scientific™
LLSR48 |
Aluminum Rod, 48 in. |
Each for $15.50
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S43741
|
United Scientific™
LLSR72 |
Aluminum Rod, 72 in. |
Each for $28.50
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Components are available individually so sets can be expanded.
- Designed to accommodate customized lattice lab setups.
- Aluminum connector used to attach two aluminum rods together.
- 12.9 mm inner diameter.
- Includes two set screws and an allen key.
- Various lengths of lattice rods available.
- Footplates available
- Hook connector available.
Specifications
Aluminum |