Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carolina™ Latex Injection Medium
For use in science education laboratories. Carolina™ Latex Injection Medium is laboratory grade injection medium in ammonia solution.
$116.00
Specifications
Chemical Name or Material | Ammonium hydroxide (>10%) |
---|---|
CAS | 1336-21-6 |
Physical Form | Liquid |
Molecular Formula | H5NO |
MDL Number | MFCD00066650 |
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Description
- No certificate of analysis is available
- Health Risk: 1
- Flammability: 0
- Reactivity: 1
Specifications
Ammonium hydroxide (>10%) | |
Liquid | |
MFCD00066650 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
amine hydrate | |
Plastic bottle |
1336-21-6 | |
H5NO | |
4 L | |
N.O | |
35.05 | |
0.96 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title