Learn More
Horiba Instruments LAQUATWIN PH-11
Supplier: Horiba Instruments 3999960122
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
LAQUAtwin pH-11 (pH meter, pH/mV, ±0.1 pH, 2 point Calibration). HORIBA INSTRUMENTS INC mfg# 3999960122
This item is not returnable.
View return policy