Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lansoprazole, Tocris Bioscience™
Click to view available options
Quantity:
50mg
Description
H+,K+-ATPase inhibitor (IC50 = 6.3 μM) that displays antisecretory and antiulcer activity. Inhibits gastric acid secretion (IC50 = 0.09 μM for histamine-induced acid formation) and reduces gastric lesion formation induced by a variety of ulcerative stimuli. Antibacterial against Helicobacter pylori in vitro. Also blocks swelling-dependent chloride channel (ICIswell) in NIH3T3 fibroblasts. More potent than omeprazole (Cat. No. 2583).
Specifications
Quantity | 50mg |
Formula Weight | 369.36 |
Percent Purity | >99% |
Chemical Name or Material | Lansoprazole |
Safety and Handling
Recommended Storage : Store at +4°C