Learn More
PATTERSON DENTAL COMPANY Contemporary Ortho-Jet, 1 lb Bottle of Powder
Supplier: PATTERSON DENTAL COMPANY 4594073
Contemporary Ortho-Jet Self Curing Acrylic Resin for Fabrication of Orthodontic Appliances, Powder, Black, 1-lb Bottle of Black Powder Only (To be used with Clear Ortho-Jet Liquid to make opaque applications)