Learn More
Getinge USA LancerClean Detergent
Supplier:  Getinge USA LCDP 2.5
Encompass
LancerClean Detergent is a concentrated, low-foaming, neutral pH detergent for use in automated endoscope reprocessors. It contains a blend of surfactants and enzymes formulated to thoroughly clean flexible endoscopes and accessories. The detergent is effective on all types of flexible endoscope materials and helps provide high-level disinfection. LancerClean Detergent is compatible with most automated endoscope reprocessors and should be diluted prior to use per the manufacturer's instructions. The product offers effective cleaning power while protecting your endoscope investment during reprocessing.
Catalog No. NC2152250
Explore available promotions