Learn More
Getinge USA Lanceracid Rinse, Acidic based, 1x5Gal
Supplier: Getinge USA LCA5A
Our most commonly used rinsing agent; LCA-A is used in the rinse cycle subsequent to our LANCERCLEAN Detergents. This acetic acid based neutralizer provides an excellent rinse on all standard laboratory soils and contaminants. LCA-A is recommended for use in all glassware washers.