Description
- Ammocoetes, larval lamprey, are filter feeders that have many anatomical features in common with Amphioxus
- Adult form develops more advanced vertebrate characteristics
Specifications
Lamprey | |
Plain | |
35.6 to 71 cm |
Formaldehyde | |
14 to 28 in. | |
10/Pk. |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov