Learn More
Cole-Parmer Lighting Specialties TEL-AF-3191 Infrared Heating Lamp, 115 V, 50/60 Hz
Supplier:  Cole-Parmer 0305700
Encompass_Preferred
Mount to any table or benchtop, or use our floor stands to support the heating lamp—features durable all-metal construction. Adjust the 42" swiveling extension arm to position lamp exactly where you want it. Dual reflector prevents excessive heat buildup. Lamp comes complete with screw-type mounting bracket; 10-ft grounded cord with plug; and 250 watt, hard glass infrared heat bulb. Easily remove wire bulb guard to replace bulb(order replacement bulb separately).
Catalog No. NC9598659