Learn More
Air Science Usa Purair FLOW Vertical Laminar Flow Cabinet, 24" Wide
SDP
Supplier:  Air Science Usa FLOW24
Encompass_Preferred
Purair FLOW Vertical Laminar Flow Cabinet (FLOW24), 24" / 600mm Nominal Width, 115V 60Hz (other voltages available). Purair FLOW Series Vertical Laminar Flow Hoods / Clean Bench are designed to protect the work surface, products and materials from particulate contamination. Room air passes through a HEPA filter then uniformly by vertical laminar flow through the cabinet interior to protect the work from unfiltered air. The airflow is oriented to exhaust airborne particulate introduced by the user. The Purair FLOW series product line employs the Air Science exclusive Multiplex HEPA filtration technology to sustain the contamination-free environment.
Catalog No. NC0771693