Learn More
Lonza Walkersville LAL Reagent Water
Supplier: Lonza Walkersville W50100
UPDATE (Mar 2018):Beginning in late March, our 100 ml (P/N W50-100), 500 ml (P/N W50-500) and 1000 ml (P/N W50-1000) LAL Reagent Water will be produced by a qualified contract manufacturer in addition to in-house manufacturing. This change is being made in order to ensure availability and maintain security of supply for these products. The contract manufacturer has been qualified by Lonza internal procedures and vendor audit. The lots sourced from the contract manufacturer will meet the release criterion that has been previously established for LAL Reagent Water produced in-house. We are therefore confident that this change will have no impact to the quality of the product.Benefits:Certified to contain less than 0.005 EU/ml endotoxinContent:1 bottle
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.