Learn More
United Scientific™ Laboratory Jack
Easily adjust with the simple turn of a knob. The Laboratory Jack is constructed of a durable combination of chemically resistant anodized aluminum and stainless steel.
Supplier: United Scientific™ LBJK01
Alert:
Prop 65 Warning
Description
- Constructed of chemically resistant anodized aluminum and stainless steel.
- The scissor jack design enables height adjustment with a simple turn of the knob.
Specifications
Anodized Aluminum, Stainless Steel | |
Closed: 2.75 in., Maximum: 10.5 in. | |
4.5 in. | |
5.5 in. | |
20 kg | |
Laboratory Jack |
17.8 cm (Closed), 26.7 cm (Maximum) | |
114.3 x 139.7 mm (4.5 x 5.5 in.) | |
114.3 mm | |
139.7 mm | |
44 lb. |