Learn More
Cole-Parmer Cole-Parmer LabGEN 125 Homogenizer Kit, Hard tissue version
Supplier:  Cole-Parmer 0472711
Encompass
Cole-Parmer’s LabGEN homogenizers are well-suited for a wide range of processing applications —use as a handheld unit or post-mount to the optional stand for hands-free operation. Plastic tip probes are maintenance-free and do not require the disassembly associated with traditional stainless steel generators. Tips are available in both soft tissue (deaggregations) and hard tissue (frozen) versions; designed to work with sample volumes between 0.25 to 30 mL. Engineered from durable plastics, these tips may be autoclaved and reused, or disposed of after each use to eliminate cross contamination.
Catalog No. NC1427783
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.