Learn More
Mirus Bio™ Label IT™ Nucleic Acid Labeling Kit, MFP488
Label IT™ Nucleic Acid Labeling Reagents for efficient, direct, non-enzymatic labeling of DNA and RNA.
$417.00 - $1005.00
Specifications
Product Type | Label Reagent |
---|---|
Concentration | 1X |
Product Line | Molecular Probes™, Click-iT™ |
Catalog Number | Mfr. No. | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Price | Quantity & Availability | |||||
MIR7100
|
Mirus Bio™
MIR 7100 |
Labeling 100 μg nucleic acid |
Each for $1,005.00
|
|
|||||
MIR7125
|
Mirus Bio™
MIR 7125 |
Labeling 25 μg nucleic acid |
Each for $417.00
|
|
|||||
Description
- Label Any DNA or RNA Template – Suitable for a wide range of applications
- One-step Chemical Method – Easily and consistently control the labeling reactions
- Adjustable Labeling Density – Achieve high sensitivity with optimally labeled DNA and RNA
- Covalent Mechanism – Permanent, non-destructive modification of nucleic acid residues is ideal for many diverse applications; labels do not impact hybridization performance
NOTE: All the Label IT Reagents use the same technology, but each attach different fluorophores with distinct excitation and emission spectra.
- MFP488 Nucleic Acid Labeling Reagent
- High Photostability – Facilitates longer time for observation and image capture
- Widespread Instrument Compatibility – Spectrally similar to fluorescein
- Stable at Low pH – Fluorescence is maintained over a broad range (pH 4-9)
Specifications
Label Reagent | |
1X | |
Molecular Probes™, Click-iT™ |