Learn More
Labconco™ Replacement Inlet or Outlet HEPA Filter
SDP
99.99% efficient on 0.3μm particles
Supplier:  Labconco™ 5225105
Catalog No. 16-108-407
Add to Cart
Description
Description
For use with Precise™ HEPA-Filtered Glove Box or XPert™ Weigh Box.
Specifications
Specifications
0.99
Precise™ HEPA Filtered Glove Box, XPert™ Weigh Box
2.9 Kg
Particulate
HEPA
6.5 lb.
Filter
Product Suggestions
Product Suggestions