Learn More
Labconco™ Purifier™ Type B2 Cabinet Remote Blower
Coated steel blower for low to moderately corrosive applications.
Supplier: Labconco™ 7071900
Description
Labconco Coated Steel Blowers have been specifically designed to meet the demanding day-to-day ventilation requirements for a wide range of hood choices. Durable and easy to maintain, Coated Steel Blowers are the recommended accessory for fume hoods used for low to moderately corrosive applications. Traditional belt driven motor between 1000 and 2000 CFM. 12.25 in. Inlet; 7.0 in. d x 13.5 in. w Outlet. 828 to 1173 RPM range available.Specifications
1 hp | |
13.5 in. | |
Coated Steel | |
Low to Moderately Corrosive Applications |
Blower | |
230 V | |
34.29 cm |