Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ PuriCare™ Bedding Disposal Station
Convenient, mobile system for disposing of used small animal bedding
Supplier: Labconco™ 3840000
Includes: 10 ft. (3m) 3-wire cord and plug
Description
- Nominal inflow velocity of 50 to 80fpm (15.2 to 24.4m/min.)
- 99.99%-efficient HEPA filter
- Rear baffle optimizes airflow and distributes air evenly across the entire work area
- Interior panel protects prefilter and HEPA filter from contact with animal cages placed inside the enclosure
- Top-mounted prefilter adsorbs odors, low molecular weight contaminants, and other gases from the exhaust airflow; also extend HEPA filter life
- Variable-speed blowers with solid-state control
- Fits easily through doorways and aisles; large, nonmarking polyurethane casters can lock in position
- Large access opening accommodates larger cages
- Large LDPE waste container on casters holds 40 gal. standard-size trash bags
- Glacier white powder-coated steel exterior
- Type 304 stainless-steel interior sides, back, upper panel, rear baffle and work surface
- 20w fluorescent light
- Work surface features 12 in. (30.5cm) square receptacle with removable trim ring
Order Info
Exterior L x W x H: 35 x 36 x 75 1/2 in.H (89 x 91 x 192cm). Interior D x W x H: 28 x 34 1/2 x 21 1/2 in. (71 x 88 x 55cm). Shipping weight: 400 lb. (182kg). Replacement waste container (09-039-122), exhaust filters (09-039-127), and dolly (09-039-123) available separately. For 115V 60Hz.
Certifications
ETL listed
Warranty and Services
Limited three-year on parts and service
Specifications
Powder | |
Fluorescent | |
5 A | |
28 in. | |
Disposing of used small animal bedding | |
21.5 in. | |
60 Hz | |
115 V | |
400 lb. | |
36 in. | |
91 cm |
HEPA | |
Powder-coated Steel, Stainless Steel (304) | |
ETL | |
71 cm | |
74.8 in. | |
190 cm | |
10 ft. 3-wire cord and plug | |
3 Years | |
182 kg | |
34.5 in. | |
88 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction