Learn More
Labconco™ Protector XVS™ Ventilation Stations: PVC Duct
Accessories for Labconco Protector XVS Ventilation Stations
Supplier: Labconco™ 4708600
Description
For use with Labconco Protector XVS Ventilation Stations
Ductwork
- Standard 6 in. dia. (15.2cm) Type I PVC duct can be cut without special tools; secured with solvent cement (Female duct coupling required to join two sections. See Cat. No. 16-304-12)
- 90° elbow (Cat. No. 16-304-11) has belled end connections to receive PVC duct directly
- Flexible duct connection reduces vibration between blower and PVC ductwork; supplied with two installation clamps
- Powder-coated steel blower transition adapter allows round PVC duct to be connected to the exhaust side of the blower
- PVC zero-pressure weathercap (Cat. No. 16-304-14) connects directly to the duct for vertical discharge of effluent air and dispersion away from the building
- Cap keeps out rain and snow but adds little static pressure to the exhaust system
Specifications
Protector XVS Ventilation Station | |
11.3 Kg | |
PVC |
25 lb. | |
PVC Duct |