Learn More
Labconco™ Protector XVS™ Blower Transition Adapter for Ventilation Stations
Blower Transition Adapter for Ventilation Stations
Supplier: Labconco™ 4861700
Description
- For use with Labconco Protector XVS Ventilation Stations
- Powder-coated steel blower transition adapter allows round PVC duct to be connected to the exhaust side of the blower
Specifications
4.25 x 7.75 in. round duct, Protector™ VS ventilation station | |
1.4 Kg | |
Epoxy-coated Steel |
3 lb. | |
Blower Transition Adapter |