Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Silver Polish for Digital Chloridometer
SDP

For use with Labconco Digital Chloridometer.

Supplier:  Labconco™ 4425094

Catalog No. 13-874-263


Add to Cart

Description

Description

  • Quantity: 0.2kg
  • Packaging: Jar
Specifications

Specifications

Digital chloridometer
8 oz.
0.2 kg
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit