Learn More
Labconco™ CentriVap™ -105 Cold Traps
SDP
Collects evaporated liquid from a CentriVap Concentrator, protecting an accessory vacuum pump from very low freezing point liquids
Supplier:  Labconco™ 7385037
Request Bulk Order
 View more versions of this product
Catalog No. 16-109-403
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
Description
Collects evaporated liquid from a CentriVap Concentrator, protecting an accessory vacuum pump from very low freezing point liquids
  • Brushed, stainless steel and glacier white, epoxy-coated steel exterior with blue accents;
  • Temperature graph illuminates green when collector coil reaches operating temperature of -65°C
  • HCFC-free refrigeration system with two 1/3 hp compressors lowers cold trap to -105°C (-157°F) in less than 40 min.
  • 4L stainless steel collector
  • Arylic lid with two Quick-Disconnect fittings for connection to a CentriVap Concentrator
  • Stainless steel drain port with corrosion-resistant, nylon valve
  • Grounding clip and ON/OFF power switch
Specifications
Specifications
CE
Benchtop Cold Trap
China/Australia
24.1 in.
17.6 in.
23.9 in.
170 lb.
230 V 50 Hz
50 Hz
Stainless Steel
Very low freezing point solvent
-105°C
61.2 cm
44.7 cm
60.7 cm
77 kg
230 V
Product Suggestions
Product Suggestions