Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lab Vision™ Tris Buffered Saline and Tween 20 (20X), Epredia™
Take advantage of a wash buffer for immunolabeling techniques with the Utilize Epredia™ Lab Vision™ Tris-Buffered Saline and Tween™ 20 (20X).
$93.00 - $552.00
Specifications
Product Type | TBS with Tween (TBST) |
---|---|
For Use With (Application) | Anatomical Pathology |
Concentration | 20X |
Content And Storage | Room Temperature |
Description
- Tween 20 is added to promote more effective washings, resulting in decreased nonspecific background staining
- Tween 20 aids reagent spreading on slides
Storage Conditions: Store at room temperature. Stable for 2 years from date of manufacture. Use diluted solution within one week
Specifications
TBS with Tween (TBST) | |
Anatomical Pathology | |
20X | |
Room Temperature |
For in vitro diagnostic use
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title