Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Pepsin Solution

Take advantage of a quality product for immunohistochemical procedures with the Epredia™ Lab Vision™ Pepsin Solution.

Supplier:  Epredia™ TA125PE

Catalog No. TA125PE

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »


Description

Description

  • pH 2.0
  • Single-component, ready-to-use format

Storage Conditions: Store at 2°-8°C. Stable for 18 months.
Specifications

Specifications

125 mL
Pepsin
2
2° to 8°C.
Immunohistochemistry
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit