Learn More
Research Products International Corp Laboratory Gravity Convection Oven, Hydraulic Thermostat, 1200 Watts
Supplier: Research Products International Corp 30200
Designed for years of dependable service. Cabinet exteriors are constructed of heavy steel with a hard scratch resistant hammer finish. Aluminized steel interiors are corrosion resistant. Energy efficient heating elements are designed for quick run-up and recovery times and provide excellent temperature uniformity. The gravity convection ovens are designed to meet the laboratory needs of industry, research organizations and schools. Well-crafted and versatile, they are used for part drying, baking, curing, sterilizing, evaporating, heat treating, annealing and testing. Heating methods: GravityThermostat Type: Hydraulic; Electrical Requirements: 120 Volts, 10.5 AMPS, 1200 Watts; Interior Dimensions: 18 x 15.6 x 12 Inches; Exterior Dimensions: 20 x 25.5 x 14.3 Inches; Capacity: 2.0 Cubic Feet; Temperature Maximum: 232°C;Shelves (1" Centers): 16 Maximum per UnitWeight (Stand Alone): 57.2 Pounds
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.