Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific Supplies Lab Clean 70% IPA Wipes
General purpose disposable 70% Isopropyl Alcohol (IPA) wipes are ideal for a variety of cleaning applications, including electronics, tools, workspaces, and other surfaces that require quick removal of contaminants. Suitable for Class 6-7 Cleanrooms.
Supplier: United Scientific Supplies ISOWP18
This item is not returnable.
View return policy
Description
- General purpose disposable 70% Isopropyl Alcohol (IPA) wipes
- Ideal for a variety of cleaning applications, including electronics, tools, workspaces, and other surfaces that require quick removal of contaminants
- Suitable for Class 6-7 Cleanrooms
- 18 cm x 14 cm (7 x 5.5 in.) perforated wipes are constructed of durable, lint-free, tear-resistant polyester
- Suitable for use in Class 6-7 Cleanrooms.
- Wipes are presaturated in a solution of 70% Isopropyl Alcohol (CAS: 63-67-0) and 30% Deionized Water
- Foil-sealed canister contains 100 wipes
- Canister includes a screw-top and a convenient flip lid with pull-through dispenser
Specifications
Alcohol Wipes | |
18 cm | |
14 cm | |
18 x 14 cm (7 x 5.5 in.) | |
70% Isoproponol Alcohol/30% Deionized Water |
7 in. | |
5.5 in. | |
Polyester, Viscose | |
100 Wipes/Canister | |
Non-sterile |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction