Learn More
Lab-Aire™ II Labware Non-Electric Benchtop Glassware Drying Racks
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
For space- and budget-conscious labs.
Supplier: Bel-Art Products F189330021/EMD
Description
Movable V-channel pegs can be repositioned to hold a variety of labware, including beakers, flasks, bottles, test tubes and cylinders.
- High storage capacity: up to 114 peg positions Single-sided benchtop models fit against a wall, conserving bench space
- Double-sided benchtop models provide double the capacity with a slightly larger footprint
- Wall-mount model has mounting points sized to fit standard pegboards to save counter space
Specifications
Benchtop Rack (1-tier) | |
38 | |
14.75 in. | |
8.4 in. | |
25.4 cm |
ABS | |
37.5 cm | |
21.3 cm | |
10 in. |