Learn More
Lab-Aids™ Blood Smear Kit
SCI_ED
Observe and identify different cell types in student's own blood, prepare smear and apply Wright's stain and develop microscope skills.
Supplier:  Lab-Aids™ 1B
Catalog No. S19330
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Requires : Microscopes
Description
Description
Lab-Aids™ blood smear three-step experiment provides students with a clear understanding of lab techniques and an opportunity for self discovery. Students draw samples of their own blood with individualized sterile lancets and prepare smears by applying Wright  s stain. Using a microscope, they examine the smears to identify different cell types (erythrocytes, polymorphonuclears, eosinophiles, basophiles, lymphocytes and platelets). This experiment can be safely conducted in a single class period. Microscopes are needed. Accommodates 2 classes, each with 25 groups of 2 students.
Specifications
Specifications
Observe and identify different cell types in their own blood, to prepare smear and apply wright's stain, to develop microscope skills
Blood Smear