Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Tryptophan methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 218°C to 220°C |
Beilstein | 4240280 |
Formula Weight | 254.71 |
Percent Purity | 98% |
Quantity | 25 g |
Chemical Name or Material | L-Tryptophan methyl ester hydrochloride |
Safety and Handling
EINECSNumber : 231-385-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only