Learn More
L-Tryptophan methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 218°C to 220°C |
Beilstein | 4240280 |
Formula Weight | 254.71 |
Percent Purity | 98% |
Quantity | 5 g |
Chemical Name or Material | L-Tryptophan methyl ester hydrochloride |
Safety and Handling
EINECSNumber : 231-385-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.