Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-Tartaric Acid, GR, ACS,, MilliporeSigma™

Catalog Number MTX00106
Click to view available options
Quantity:
5 kg
This item is not returnable. View return policy
87-69-4
C4H6O6
150.09
MFCD00064207
FEWJPZIEWOKRBE-UHFFFAOYNA-N
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
444305
CHEBI:15671
2,3-dihydroxybutanedioic acid
OC(C(O)C(O)=O)C(O)=O
This item is not returnable. View return policy
87-69-4
C4H6O6
150.09
MFCD00064207
FEWJPZIEWOKRBE-UHFFFAOYNA-N
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
444305
CHEBI:15671
2,3-dihydroxybutanedioic acid
OC(C(O)C(O)=O)C(O)=O
Color White
Formula Weight 150.09
Quantity 5 kg
Physical Form Solid
Chemical Name or Material L-(+)-Tartaric Acid, For ACS analysis
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit