Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Lactic Acid (Free Acid), MP Biomedicals™
Click to view available options
Quantity:
1 g
25 g
Specifications
Color | White |
Formula Weight | 90.08 |
Density | 1190 to 1250kg/mL |
Quantity | 25 g |
Physical Form | Solid |
Chemical Name or Material | L-Lactic Acid, Free Acid |
Safety and Handling
Recommended Storage | Store at 0°C. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction