Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ L-Histidine, Molecular biology grade, OmniPur™, Calbiochem™,

Click to view available options
Quantity:
5 kg
Packaging:
Plastic Drum
Color White
Packaging Plastic Drum
Percent Purity 98.5 to 101.5%
Grade Molecular Biology
Quantity 5 kg
Physical Form Solid