Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mirus Bio™ L Glutamine Solution, 200 mM
L-Glutamine is an essential amino acid that is added as a carbon and energy source to mammalian and insect cell culture media.
Supplier: Mirus Bio™ MIR 6240
This item is not returnable.
View return policy
Description
- L-Glutamine is an essential amino acid that is added as a carbon and energy source to mammalian and insect cell culture media.
- Optimal cell performance usually requires 2-4 mM L-Glutamine supplementation.
- L-Glutamine gradually degrades in stock solutions and liquid media and should be added immediately prior to use.
- L-Glutamine, 200 mM Solution (MIR 6240) is a sterile-filtered, animal-origin-free reagent.
Specifications
56-85-9 | |
Store at -20°C | |
L-Glutamine | |
0.2 μm Sterile-filtered | |
None |
Colorless | |
Liquid | |
Sterile | |
None | |
100 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction