Learn More
Pfanstiehl Inc L-Glutamine, High Purity, Low Endotoxin, Low Metals, USP, JP, ChP (G-137) - 5kg pack GMP, Multicompendial -CAS: 56-85-9
Supplier:  Pfanstiehl Inc G1375KG
Encompass_Preferred
L-Glutamine is an essential amino acid involved in numerous physiological processes within mammalian cells. In upstream cell culture protein processes, L-Glutamine plays a pivotal role in supporting cell growth, viability, and protein production.
Catalog No. NC2271933