Learn More
Bellco Glass, Inc. Manufactured from 316l stainless steel with electropolish finish. Ra 32 finish or better inside and out. Ideal use for cell culture and reduces shear in delicate cell culture. Blade dimension: 5 3/4 x 2 1/2" Shaft oal: 12" Marine impeller incl
Supplier: Bellco Glass, Inc. 196488088
Manufactured from 316l stainless steel with electropolish finish. Ra 32 finish or better inside and out. Ideal use for cell culture and reduces shear in delicate cell culture. Blade dimension: 5 3/4 x 2 1/2" Shaft oal: 12" Marine impeller incl